Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036347.2 | NP_001031424.1 | 2 - 3 | 400 | 72 | |
NM_001036348.2 | NP_001031425.1 | 3 - 4 | 696 | 72 | ||
NM_001336077.1 | NP_001325357.1 | 3 - 4 | 233 | 72 | ||
NM_128216.3 | NP_565626.1 | 3 - 4 | 661 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |