Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336091.1 | NP_001324382.1 | 4 - 5 | 911 | 82 | |
NM_001336092.1 | NP_001324381.1 | 4 - 5 | 899 | 82 | ||
NM_001336093.1 | NP_001324383.1 | 4 - 5 | 899 | 82 | ||
NM_128239.1 | NP_180250.1 | 4 - 5 | 792 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |