Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPMEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128250.4 | NP_180261.1 | 8 - 9 | 930 | 79 | |
GenBank mRNA | AB037877.1 | - | 8 - 9 | 874 | 79 | |
AX765854.1 | - | 8 - 9 | 869 | 79 | ||
AY136394.1 | - | 8 - 9 | 886 | 79 | ||
BT000208.1 | - | 8 - 9 | 869 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |