Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336097.1 | NP_001323455.1 | 3 - 4 | 547 | 77 | |
NM_128260.5 | NP_180270.1 | 3 - 4 | 393 | 77 | ||
GenBank mRNA | AF043528.1 | - | 3 - 4 | 270 | 77 | |
AF375455.1 | - | 3 - 4 | 260 | 77 | ||
AY088609.1 | - | 3 - 4 | 348 | 77 | ||
AY124806.1 | - | 3 - 4 | 227 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |