Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336102.1 | NP_001325161.1 | 44 - 45 | 5540 | 111 | |
NM_001336103.1 | NP_001325160.1 | 44 - 45 | 5534 | 111 | ||
NM_128270.3 | NP_180280.2 | 44 - 45 | 5531 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |