Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036353.1 | NP_001031430.1 | 2 - 3 | 544 | 90 | |
NM_128281.2 | NP_565640.1 | 2 - 3 | 507 | 90 | ||
GenBank mRNA | AB493566.1 | - | 1 - 2 | 95 | 90 | |
AJ576043.1 | - | 1 - 2 | 95 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |