Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336138.1 | NP_001324957.1 | 3 - 4 | 1688 | 60 | |
NM_128358.3 | NP_180366.1 | 4 - 5 | 1592 | 60 | ||
GenBank mRNA | AK119153.1 | - | 4 - 5 | 1542 | 60 | |
BT005921.1 | - | 3 - 4 | 1414 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |