Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336157.1 | NP_001324840.1 | 1 - 2 | 693 | 140 | |
NM_128386.2 | NP_180393.1 | 2 - 3 | 1093 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |