Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036361.2 | NP_001031438.1 | 4 - 5 | 505 | 89 | |
NM_001124931.1 | NP_001118403.1 | 3 - 4 | 416 | 89 | ||
NM_001161066.1 | NP_001154538.1 | 3 - 4 | 567 | 89 | ||
NM_001336172.1 | NP_001323470.1 | 4 - 5 | 506 | 89 | ||
NM_001336173.1 | NP_001323469.1 | 4 - 5 | 505 | 89 | ||
NM_128416.4 | NP_180423.3 | 4 - 5 | 512 | 89 | ||
GenBank mRNA | AB194809.1 | - | 4 - 5 | 490 | 89 | |
AB194810.1 | - | 3 - 4 | 567 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |