Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036361.2 | NP_001031438.1 | 11 - 12 | 1127 | 70 | |
NM_001124931.1 | NP_001118403.1 | 10 - 11 | 1038 | 70 | ||
NM_001336172.1 | NP_001323470.1 | 11 - 12 | 1128 | 70 | ||
NM_001336173.1 | NP_001323469.1 | 11 - 12 | 1127 | 70 | ||
GenBank mRNA | AB194809.1 | - | 11 - 12 | 1112 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |