Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTU5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128499.5 | NP_180506.1 | 1 - 2 | 540 | 108 | |
GenBank mRNA | AF144382.1 | - | 1 - 2 | 362 | 108 | |
AY062676.1 | - | 1 - 2 | 383 | 108 | ||
AY088413.1 | - | 1 - 2 | 385 | 108 | ||
BT001228.1 | - | 1 - 2 | 316 | 108 | ||
D44465.1 | - | 1 - 2 | 372 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |