Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336222.1 | NP_001324956.1 | 8 - 9 | 1451 | 149 | |
NM_128523.5 | NP_180530.1 | 8 - 9 | 1496 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |