Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113013.5 | NP_188755.2 | 4 - 5 | 887 | 137 | |
GenBank mRNA | AK227228.1 | - | 4 - 5 | 844 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |