Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113046.4 | NP_188788.1 | 4 - 5 | 582 | 120 | |
GenBank mRNA | AB012570.1 | - | 4 - 5 | 364 | 120 | |
AB015141.1 | - | 4 - 5 | 370 | 120 | ||
AF370265.1 | - | 4 - 5 | 379 | 120 | ||
AY063082.1 | - | 4 - 5 | 330 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |