Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT71B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113073.3 | NP_188815.2 | 1 | 694 | 76 | |
GenBank mRNA | KJ138815.1 | - | 1 | 665 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |