Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113082.4 | NP_566696.2 | 1 - 2 | 403 | 63 | |
GenBank mRNA | AY063961.1 | - | 1 - 2 | 228 | 63 | |
AY096402.1 | - | 1 - 2 | 117 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |