Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338564.1 | NP_001327425.1 | 1 | 1203 | 123 | |
NM_113109.3 | NP_188851.2 | 1 | 1053 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |