Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203018.1 | NP_001189947.1 | 2 - 3 | 488 | 67 | |
NM_113117.4 | NP_566700.1 | 2 - 3 | 285 | 67 | ||
GenBank mRNA | AY034923.1 | - | 2 - 3 | 276 | 67 | |
AY142571.1 | - | 2 - 3 | 148 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |