Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125204.1 | NP_001118676.1 | 1 - 2 | 372 | 69 | |
NM_001338582.1 | NP_001326717.1 | 1 - 2 | 372 | 69 | ||
NM_113136.4 | NP_566705.1 | 1 - 2 | 372 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |