Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113173.3 | NP_566715.1 | 2 - 3 | 533 | 104 | |
GenBank mRNA | AF297394.1 | - | 2 - 3 | 238 | 104 | |
AK118021.1 | - | 2 - 3 | 333 | 104 | ||
BT005318.1 | - | 2 - 3 | 238 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |