Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338596.1 | NP_001325669.1 | 4 - 5 | 1130 | 122 | |
NM_113175.2 | NP_566717.1 | 4 - 5 | 1142 | 122 | ||
GenBank mRNA | AY099653.1 | - | 4 - 5 | 1013 | 122 | |
AY128837.1 | - | 4 - 5 | 947 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |