Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KINESIN-12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035673.2 | NP_001030750.1 | 8 - 9 | 1062 | 105 | |
NM_113271.4 | NP_189009.2 | 8 - 9 | 1092 | 105 | ||
GenBank mRNA | AF540489.1 | - | 8 - 9 | 1064 | 105 | |
AY124006.1 | - | 8 - 9 | 1060 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |