Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALFL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113285.3 | NP_566740.1 | 1 | 473 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |