Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113306.4 | NP_566742.1 | 5 - 6 | 538 | 76 | |
GenBank mRNA | AB493630.1 | - | 5 - 6 | 438 | 76 | |
BT024927.1 | - | 5 - 6 | 438 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |