Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113321.5 | NP_189058.1 | 4 - 5 | 484 | 87 | |
GenBank mRNA | AF104329.1 | - | 4 - 5 | 348 | 87 | |
AY053413.1 | - | 4 - 5 | 364 | 87 | ||
AY085022.1 | - | 4 - 5 | 384 | 87 | ||
AY133587.1 | - | 4 - 5 | 299 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |