Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113359.4 | NP_566749.1 | 4 - 5 | 989 | 75 | |
GenBank mRNA | AF349448.1 | - | 4 - 5 | 754 | 75 | |
AY056398.1 | - | 4 - 5 | 760 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |