Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PERK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113366.3 | NP_189098.1 | 3 - 4 | 1502 | 79 | |
GenBank mRNA | AY056788.1 | - | 3 - 4 | 1222 | 79 | |
AY059901.1 | - | 3 - 4 | 1217 | 79 | ||
AY093065.1 | - | 3 - 4 | 1215 | 79 | ||
AY128792.1 | - | 3 - 4 | 1186 | 79 | ||
BT008400.1 | - | 3 - 4 | 1186 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |