Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113392.3 | NP_189124.1 | 7 - 8 | 1089 | 68 | |
GenBank mRNA | AJ224307.1 | - | 7 - 8 | 1066 | 68 | |
AY080799.1 | - | 7 - 8 | 1087 | 68 | ||
AY114077.1 | - | 7 - 8 | 945 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |