Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203042.1 | NP_001189971.1 | 8 - 9 | 1154 | 154 | |
NM_113453.3 | NP_189183.2 | 8 - 9 | 1200 | 154 | ||
NM_202631.2 | NP_974360.1 | 8 - 9 | 985 | 154 | ||
GenBank mRNA | AY075617.1 | - | 8 - 9 | 933 | 154 | |
BT002619.1 | - | 8 - 9 | 854 | 154 | ||
FJ708728.1 | - | 8 - 9 | 854 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |