Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203042.1 | NP_001189971.1 | 9 - 10 | 1492 | 69 | |
NM_113453.3 | NP_189183.2 | 9 - 10 | 1538 | 69 | ||
NM_202631.2 | NP_974360.1 | 9 - 10 | 1323 | 69 | ||
GenBank mRNA | AY075617.1 | - | 9 - 10 | 1271 | 69 | |
BT002619.1 | - | 9 - 10 | 1192 | 69 | ||
FJ708728.1 | - | 9 - 10 | 1192 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |