Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113607.4 | NP_566804.3 | 5 - 6 | 2031 | 106 | |
GenBank mRNA | AF002706.1 | - | 5 - 6 | 1913 | 106 | |
AF026549.1 | - | 5 - 6 | 1893 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |