Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLC-B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113631.4 | NP_189353.1 | 2 - 3 | 587 | 65 | |
GenBank mRNA | AY062518.1 | - | 2 - 3 | 501 | 65 | |
Z71446.1 | - | 2 - 3 | 456 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |