Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203056.2 | NP_001189985.1 | 6 - 7 | 1934 | 131 | |
NM_001338864.1 | NP_001327404.1 | 6 - 7 | 2026 | 131 | ||
NM_113640.2 | NP_189362.1 | 6 - 7 | 2010 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |