Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035703.3 | NP_001030780.1 | 7 - 8 | 946 | 117 | |
NM_001084745.1 | NP_001078214.1 | 6 - 7 | 772 | 117 | ||
NM_001338866.1 | NP_001325679.1 | 6 - 7 | 909 | 117 | ||
NM_001338867.1 | NP_001325680.1 | 6 - 7 | 919 | 117 | ||
NM_113644.5 | NP_189366.1 | 6 - 7 | 869 | 117 | ||
GenBank mRNA | AK317166.1 | - | 7 - 8 | 841 | 117 | |
BT021101.1 | - | 5 - 6 | 678 | 117 | ||
BT029298.1 | - | 5 - 6 | 672 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |