Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113645.3 | NP_566815.1 | 4 - 5 | 504 | 65 | |
GenBank mRNA | AK175284.1 | - | 4 - 5 | 333 | 65 | |
AY572781.1 | - | 4 - 5 | 321 | 65 | ||
BT024476.1 | - | 4 - 5 | 321 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |