Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113645.3 | NP_566815.1 | 5 - 6 | 640 | 84 | |
GenBank mRNA | AK175284.1 | - | 5 - 6 | 469 | 84 | |
AY572781.1 | - | 5 - 6 | 457 | 84 | ||
BT024476.1 | - | 5 - 6 | 457 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |