Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113645.3 | NP_566815.1 | 6 - 7 | 748 | 107 | |
GenBank mRNA | AK175284.1 | - | 6 - 7 | 577 | 107 | |
AY085053.1 | - | 6 - 7 | 589 | 107 | ||
AY572781.1 | - | 6 - 7 | 565 | 107 | ||
BT024476.1 | - | 6 - 7 | 565 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |