Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125658.3 | NP_201069.1 | 6 - 7 | 1864 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |