Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125705.4 | NP_201116.1 | 3 - 4 | 1245 | 77 | |
GenBank mRNA | AY072201.1 | - | 3 - 4 | 1200 | 77 | |
AY088314.1 | - | 3 - 4 | 1224 | 77 | ||
AY142660.1 | - | 3 - 4 | 1125 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |