Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125726.4 | NP_568970.2 | 2 - 3 | 363 | 101 | |
GenBank mRNA | AJ012758.1 | - | 2 - 3 | 297 | 101 | |
AK227190.1 | - | 2 - 3 | 318 | 101 | ||
AY057612.1 | - | 2 - 3 | 316 | 101 | ||
AY065291.1 | - | 2 - 3 | 314 | 101 | ||
AY117366.1 | - | 2 - 3 | 297 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |