Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126014.1 | NP_001119486.1 | 7 - 8 | 1800 | 120 | |
NM_125749.5 | NP_201159.2 | 6 - 7 | 1734 | 120 | ||
GenBank mRNA | AY735746.1 | - | 6 - 7 | 1699 | 120 | |
AY954880.1 | - | 6 - 7 | 1636 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |