Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125776.2 | NP_201186.1 | 16 - 17 | 2049 | 69 | |
GenBank mRNA | AY093118.1 | - | 16 - 17 | 2049 | 69 | |
BT010338.1 | - | 16 - 17 | 1881 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |