Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI-4KBETA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345618.1 | NP_001330874.1 | 3 - 4 | 2343 | 113 | |
NM_125803.4 | NP_201212.1 | 3 - 4 | 2375 | 113 | ||
GenBank mRNA | AF462854.1 | - | 3 - 4 | 2343 | 113 | |
AY139795.1 | - | 3 - 4 | 1799 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |