Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037065.2 | NP_001032142.1 | 8 - 9 | 990 | 97 | |
NM_001345648.1 | NP_001332553.1 | 7 - 8 | 1093 | 97 | ||
NM_125872.4 | NP_568994.2 | 8 - 9 | 1021 | 97 | ||
GenBank mRNA | AY062093.2 | - | 8 - 9 | 996 | 97 | |
AY062113.1 | - | 8 - 9 | 996 | 97 | ||
AY124872.1 | - | 7 - 8 | 880 | 97 | ||
AY230833.1 | - | 7 - 8 | 883 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |