Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125877.4 | NP_568995.2 | 1 - 2 | 412 | 73 | |
GenBank mRNA | AB493813.1 | - | 1 - 2 | 244 | 73 | |
AF426252.1 | - | 1 - 2 | 244 | 73 | ||
BT025755.1 | - | 1 - 2 | 244 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |