Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125884.4 | NP_201291.1 | 6 - 7 | 876 | 94 | |
GenBank mRNA | AY037231.1 | - | 6 - 7 | 805 | 94 | |
AY081744.1 | - | 6 - 7 | 785 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |