Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037068.1 | NP_001032145.1 | 3 - 4 | 1490 | 131 | |
NM_125910.6 | NP_201316.1 | 3 - 4 | 1655 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |