Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161327.1 | NP_001154799.1 | 3 - 4 | 681 | 66 | |
NM_001203697.1 | NP_001190626.1 | 3 - 4 | 957 | 66 | ||
NM_001203698.1 | NP_001190627.1 | 3 - 4 | 981 | 66 | ||
NM_125974.5 | NP_201378.5 | 3 - 4 | 681 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |