Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161327.1 | NP_001154799.1 | 6 - 7 | 2826 | 116 | |
NM_001203697.1 | NP_001190626.1 | 6 - 7 | 3021 | 116 | ||
NM_001203698.1 | NP_001190627.1 | 6 - 7 | 3045 | 116 | ||
NM_125974.5 | NP_201378.5 | 6 - 7 | 2745 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |