Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345729.1 | NP_001331804.1 | 10 - 11 | 1233 | 68 | |
NM_125994.2 | NP_201398.1 | 10 - 11 | 1324 | 68 | ||
GenBank mRNA | AF369571.1 | - | 10 - 11 | 1009 | 68 | |
BT033095.1 | - | 10 - 11 | 964 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |